Analog Micrometer

Get highly accurate measurements
  • Durable for a wide range of applications
  • Accurate to ±0.0001 in
Friction thimble for accurate readings with high repeatability; positive-locking clamp; clear graduations; zero adjustment; baked enamel finish with carbide-tipped measuring faces.

Use this micrometer for highly accurate measurements. Micrometers include case.
$131.72 - $561.73CAD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (mm)
Availability Pricing
Fowler 52-229-201 1in Economy Micrometer - +/-0.0001 in 0992040 Mfr # 522292010
COMPARE
25
$131.72CAD / Each
Fowler 52-229-213 Analog micrometer set - +/-0.0001 in (0 to 3 in/0 to 75mm) 0992094 Mfr # 522292130
COMPARE
75
$561.73CAD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat