Alicat Whisper™ MCW-Series Low-Pressure Drop Mass Flow Controllers, SLPM
Accurately measures and controls real-time mass and volumetric flow readings with minimal pressure drop
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ideal for systems with limited source pressure
  • Less than one millisecond sensor response time
  • IP40 rating
These low-pressure drop mass flow controllers, featuring a fast control response time of just 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. Controllers create minimal pressure drop, making them ideal for systems with limited source pressure. They are capable of measuring and controlling flow generated by both positive pressure and suction. Real-time monitoring of flow, pressure, and temperature is available directly on the display screen.
The controllers come with 98 pre-loaded gas calibrations, which can be seamlessly switched while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure, with flow rates displayed in standard liters per minute (SLPM). Additional key features include an LCD display with backlight, a data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
$2,961.51 - $6,497.65CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Process Connection (NPT(F)) Accuracy
Availability Pricing
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 1 SLPM 6780392 Mfr # MCW-1SLPM-D
COMPARE
1 1/8 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$2,961.51CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 2 SLPM 6780393 Mfr # MCW-2SLPM-D
COMPARE
2 1/8 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$2,961.51CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 5 SLPM 6780394 Mfr # MCRW-5SLPM-D
COMPARE
5 1/4 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$3,300.39CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 10 SLPM 6780395 Mfr # MCRW-10SLPM-D
COMPARE
10 1/4 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$3,300.39CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 20 SLPM 6780396 Mfr # MCRW-20SLPM-D
COMPARE
20 1/4 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$3,300.39CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 40 SLPM 6780397 Mfr # MCRW-40SLPM-D
COMPARE
40 1/2 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$4,213.90CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 50 SLPM 6780398 Mfr # MCRW-50SLPM-D
COMPARE
50 3/4 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$4,213.90CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 100 SLPM 6780399 Mfr # MCRW-100SLPM-D
COMPARE
100 3/4 ±0.75% of reading or ±0.1% of full scale, whichever is greater
$4,781.15CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 250 SLPM 6780400 Mfr # MCRW-250SLPM-D
COMPARE
250 3/4 ±0.8% of reading + 0.2% of full scale
$4,781.15CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flow Controller, 0 - 500 SLPM 6780401 Mfr # MCRW-500SLPM-D
COMPARE
500 3/4 ±0.8% of reading + 0.2% of full scale
$6,497.65CAD / Each
Showing 1- 10 of 10