Alicat MB-Series Mass Flowmeters with Digital Display, Battery Operated, SCCM
Provides accurate, real-time mass and volumetric flow readings
  • Built-in rechargeable battery with up to 18 hours life
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.5% of reading
  • IP40 rating
  • Limited lifetime warranty
These portable mass flowmeters, with a fast control response time of 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They come with 98 pre-loaded gas calibrations, which can be switched seamlessly while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure with flow rates displayed in standard cubic centimeters per minute (SCCM). Key features include a rechargeable battery, backlit monochrome display, data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
$2,313.22 - $3,145.69CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (SCCM) Accuracy Repeatability
Availability Pricing
Alicat Mass Flowmeter with Display, 0 - 0.5 SCCM, Battery Operated 6780269 Mfr # MB-0.5SCCM-D
COMPARE
0.5 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$3,145.69CAD / Each
Alicat Mass Flowmeter with Display, 0 - 1 SCCM, Battery Operated 6780270 Mfr # MB-1SCCM-D
COMPARE
1 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$3,145.69CAD / Each
Alicat Mass Flowmeter with Display, 0 - 2 SCCM, Battery Operated 6780271 Mfr # MB-2SCCM-D
COMPARE
2 ±0.4% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$3,145.69CAD / Each
Alicat Mass Flowmeter with Display, 0 - 5 SCCM, Battery Operated 6780272 Mfr # MB-5SCCM-D
COMPARE
5 ±0.6% of reading ±0.2% of reading + 0.02% of full scale
$2,313.22CAD / Each
Alicat Mass Flowmeter with Display, 0 - 10 SCCM, Battery Operated 6780273 Mfr # MB-10SCCM-D
COMPARE
10 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$2,313.22CAD / Each
Alicat Mass Flowmeter with Display, 0 - 20 SCCM, Battery Operated 6780274 Mfr # MB-20SCCM-D
COMPARE
20 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$2,313.22CAD / Each
Alicat Mass Flowmeter with Display, 0 - 50 SCCM, Battery Operated 6780275 Mfr # MB-50SCCM-D
COMPARE
50 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$2,313.22CAD / Each
Alicat Mass Flowmeter with Display, 0 - 100 SCCM, Battery Operated 6780276 Mfr # MB-100SCCM-D
COMPARE
100 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$2,313.22CAD / Each
Alicat Mass Flowmeter with Display, 0 - 200 SCCM, Battery Operated 6780277 Mfr # MB-200SCCM-D
COMPARE
200 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$2,313.22CAD / Each
Alicat Mass Flowmeter with Display, 0 - 500 SCCM, Battery Operated 6780278 Mfr # MB-500SCCM-D
COMPARE
500 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$2,313.22CAD / Each
Showing 1- 10 of 10