Advantec Disposable Syringe Filters, Mixed Cellulose Ester
Housings are specifically designed to minimize sample holdup and maximize recovery
  • Non-pigmented acrylic or polypropylene (PP) housings feature integral filter sealing to avoid the risk of contamination from pigments and adhesives
  • Each filter is clearly marked with membrane pore size
  • Housings have standard luer connectors; 50-mm filters feature 7 to 13.5 mm hose barb connectors
Mixed Cellulose Ester Filters are hydrophilic with high porosity and fast flow rate. A good choice for general purpose filtration of aqueous solutions. Offers higher protein binding than cellulose acetate.
$179.02 - $242.74CAD / Pkg of 50
3 variations of this product are available.
See All Product Options Below
RESOURCES
+ SAVE TO MY DOCS (PDFs)
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
Advantec 25AS020AS Syringe Filters, 0.20 um MCE, 25 mm, Sterile; 50/Pk 8105426 Mfr # 25AS020AS
COMPARE
25 0.2 Yes
$242.74CAD / Pkg of 50
Advantec 25AS045AN Syringe Filters, 0.45 um MCE, 25 mm; 50/Pk 8105428 Mfr # 25AS045AN
COMPARE
25 0.45 No
$179.02CAD / Pkg of 50
Advantec 25AS045AS Syringe Filters, 0.45 um MCE, 25 mm, Sterile; 50/Pk 8105472 Mfr # 25AS045AS
COMPARE
25 0.45 Yes
$241.64CAD / Pkg of 50