ADAM® Highland™ Portable Balances with NTEP Approval
Loaded with features for everyday weighing applications
  • Internal rechargeable battery for convenient portable operation
  • Standard draft shield offers greater weighing accuracy
  • Unique shape allows convenient stackable storage of multiple balances
  • Three-point shock protection prevents overload damage to internal components
  • NTEP-approved models (d≤0.5g e≤0.5g) available for commercial applications
Loaded with features that include multiple weighing units, RS-232 and USB interfaces, rechargeable battery pack, capacity tracker, security lock down port, and below balance weighing hook for density and specific gravity applications. A standard removable draft shield eliminates wind disturbances outdoors or in drafty environments. Backlit display with large 1/2"-high digits offers a clear view of weighing results.
NTEP-approved models feature a wide range of capacities and specifications for both the smallest (d≤0.5 g) and the verification (e≤0.5 g) scale divisions, ensuring commercial weights are easy to interpret. Select models with NTEP approval if your weighing application includes buying, selling, or charging based upon weight within the United States. These models have been tested and proven to meet all government standards and requirements and are considered legal for trade.
$1,053.47 - $1,355.52CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g)
Availability Pricing
Adam Equipment HCB 103aM Highland® Portable Balance, NTEP Approved, 100g x d=0.001g e=0.01g 1112551 Mfr # HCB 103AM
COMPARE
100 0.001
$1,326.05CAD / Each
Adam Equipment HCB 302aM Highland® Portable Balance, NTEP Approved, 300g x d=0.05g e=0.05g 1112552 Mfr # HCB 302AM
COMPARE
300 0.05
$1,053.47CAD / Each
Adam Equipment HCB 602aM Highland® Portable Balance, NTEP Approved, 600g x d=0.01g e=0.1g 1112553 Mfr # HCB 602AM
COMPARE
600 0.01
$1,163.98CAD / Each
Adam Equipment HCB 1202aM Highland® Portable Balance, NTEP Approved, 1200g x d=0.01g e=0.1g 1112554 Mfr # HCB 1202AM
COMPARE
1200 0.01
$1,355.52CAD / Each
Adam Equipment HCB 3001aM Highland® Portable Balance, NTEP Approved, 3000g x d=0.5g e=0.5g 1112555 Mfr # HCB 3001aM
COMPARE
3000 0.5
$1,134.51CAD / Each
Adam Equipment HCB 5001aM Highland® Portable Balance, NTEP Approved, 5000g x d=0.5g e=0.5g 1112556 Mfr # HCB 5001aM
COMPARE
5000 0.5
$1,163.98CAD / Each