Adam Equipment Stainless Steel Weighing Platform Scales
Durable scales serve a variety of heavy-duty needs
  • Made to handle high-volume industrial applications
  • Overload protection helps prevent damage to internal components
  • Available in a range of capacities and platform sizes
  • Can be used with any Adam indicator
A removable stainless steel pan and cast-aluminum base make these sleek yet sturdy platform scales ideal for weighing, parts counting, percentage/comparative weighing and other industrial applications. Featuring single-point precision load cell technology, the scales are well-suited for shipping and receiving, batching items for processing and stock auditing.
Work well as an auxiliary platform in tandem with the Adam CBD Series counting scales.
$1,141.88 - $1,606.00CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Readability (g)
Availability Pricing
Adam Equipment GB35a Stainless Steel Weighing Scale, 35lb/16kg, 11.8" x 15.7" Platform 1113036 Mfr # GB 35A
COMPARE
16 35 0.5
$1,141.88CAD / Each
Adam Equipment GB70a Stainless Steel Weighing Scale, 70lb/32kg, 11.8" x 15.7" Platform 1113037 Mfr # GB 70A
COMPARE
32 70 1
$1,141.88CAD / Each
Adam Equipment GB130a Stainless Steel Weighing Scale, 130lb/60kg, 11.8" x 15.7" Platform 1113038 Mfr # GB 130A
COMPARE
60 130 2
$1,141.88CAD / Each
Adam Equipment GB260a Stainless Steel Weighing Scale, 260lb/120kg, 11.8" x 15.7" Platform 1113039 Mfr # GB 260A
COMPARE
120 260 5
$1,141.88CAD / Each
Adam Equipment GF 165A Stainless Steel Weighing Scale, 165lb/75kg, 15.7" x 19.7" Platform 1113040 Mfr # GF 165A
COMPARE
75 165 5
$1,171.35CAD / Each
Adam Equipment GF330a Stainless Steel Weighing Scale, 330lb/150kg, 15.7" x 19.7" Platform 1113041 Mfr # GF 330A
COMPARE
150 330 10
$1,171.35CAD / Each
Adam Equipment GF 660A Stainless Steel Weighing Scale, 660lb/300kg, 15.7" x 19.7" Platform 1113042 Mfr # GF 660A
COMPARE
300 660 20
$1,171.35CAD / Each
Adam Equipment GF1320a Stainless Steel Weighing Scale, 1320lb/600kg, 23.6" x 31.5" Platform 1113043 Mfr # GF 1320A
COMPARE
600 1320 50
$1,606.00CAD / Each
Showing 1- 8 of 8