Adam Equipment Solis Analytical and Semi-Micro Balances
Intuitive graphic display and keypad simplifies weighing operations
  • Formulation and recipe function stores and recalls up to 99 ingredients
  • Selectable digital filtering minimizes effects of vibration and disturbances
  • Level indicator and adjustable feet ensure proper balance setup
  • Security slot for optional Kensington style lock and cable prevents theft
  • Large stainless-steel weighing pan allows quick cleaning
  • Case and draft shield reduce static electrical interference
  • Time and date stamp meets Good Laboratory Practice (GLP) guidelines
The Solis line of analytical balances include a large high-resolution graphic display where you can readily view your comprehensive data. Intuitive keypad, easy-to-read buttons, and navigation arrows simplify balancing operations. Measurements display in seconds with all pertinent data for the active application. A built-in memory stores and recall products, formulas, and settings.
The balances also feature RS-232 interface to transfer data to computers and printers, checkweighing with audible alarm, part counting with preset sample sizes, dynamic weighing for animals or liquids, percentage weighing, density determination with optional density kit, security password protection, and multilingual display capability.
Choose from a variety of capacities with external calibration (must use calibration mass to calibrate), or internal calibration (may use with or without a calibration mass).
$3,867.65 - $8,626.70CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Description
Availability Pricing
Adam Equipment SAB 125i Solis Semi-Micro Balance, 62g x .01mg / 120g x.1mg, Internal Calibration 1112528 Mfr # SAB 125I
COMPARE
Internal 120 Solis Semi-Micro Balance, 62g x .01mg / 120g x.1mg, Internal Calibration
$7,890.00CAD / Each
Adam Equipment SAB 314e Solis Analytical Balance, 310g x 0.0001g, External Calibration 1112533 Mfr # SAB 314E
COMPARE
External 310 Solis Analytical Balance, 310g x 0.0001g, External Calibration
$4,766.42CAD / Each
Adam Equipment SAB 124i Solis Analytical Balance,120g x 0.0001g, Internal Calibration 1112535 Mfr # SAB 124I
COMPARE
Internal 120 Solis Analytical Balance,120g x 0.0001g, Internal Calibration
$3,867.65CAD / Each
Adam Equipment SAB 224i Solis Analytical Balance, 220g x 0.0001g, Internal Calibration 1112536 Mfr # SAB 224I
COMPARE
Internal 220 Solis Analytical Balance, 220g x 0.0001g, Internal Calibration
$4,589.61CAD / Each
Adam Equipment SAB 414i Solis Analytical Balance, 410g x 0.0001g, Internal Calibration 1112538 Mfr # SAB 414I
COMPARE
Internal 410 Solis Analytical Balance, 410g x 0.0001g, Internal Calibration
$6,784.96CAD / Each
Adam Equipment SAB 514i Solis Analytical Balance, 510g x 0.0001g, Internal Calibration 1112539 Mfr # SAB 514I
COMPARE
Internal 510 Solis Analytical Balance, 510g x 0.0001g, Internal Calibration
$8,626.70CAD / Each