Adam Equipment SHS Crane Scales
Flexible crane scales make it easy to measure suspended loads
  • Three-button infrared remote control
  • Color-coded keypad
  • Three weighing units (kg, lb, Newtons)
  • Display hold function
  • 360-degree rotating display
  • 360-degree swivel shackle and bolt
  • External calibration
  • Polycarbonate indicator housing (SHS only)
  • Cast aluminum magnesium indicator housing (LHS only)
  • Chrome-plated steel hook with safety catch
  • Automatic power-off
  • Low-battery indicator
  • Rechargeable battery
Designed with safety in mind each scale features a standard remote control that allows operation at a safe distance. The rotating hook with safety catch ensures stability and keeps the suspended item securely in place. High-contrast red LED display and indicators are easy to read in any lighting conditions. A handy hold function keeps the reading visible on the display after the weight is removed, allowing the operator to safely record the weight.
Built-in rechargeable battery - Alows the scale to be used in areas where power is not available.
Simple, Flexible Operation - Use the included remote control or the colourcoded keypad to safely operate the scale’s functions. The bright-yellow tare/zero key is easy to spot for quick recognition.
$1,001.91 - $1,053.47CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Description
Availability Pricing
Adam Equipment SHS 100A Battery Operated Crane Scales; 50kg x 0.01kg 1171009 Mfr # SHS 100A
COMPARE
50 100 Battery Operated Crane Scales; 50kg x 0.01kg
$1,001.91CAD / Each
Adam Equipment SHS 300A Battery Operated Crane Scales; 150kg x 0.02kg 1171010 Mfr # SHS 300A
COMPARE
150 300 Battery Operated Crane Scales; 150kg x 0.02kg
$1,038.74CAD / Each
Adam Equipment SHS 600A Battery Operated Crane Scales; 300kg x 0.05kg 1171011 Mfr # SHS 600A
COMPARE
300 600 Battery Operated Crane Scales; 300kg x 0.05kg
$1,053.47CAD / Each
Adam Equipment SHS 100A Crane Scale, 50kg with 0.01kg Readability, Led, Lithium Battery 220V 1171039 Mfr # SHS 50
COMPARE
50 110.4 Crane Scale, 50kg with 0.01kg Readability, Led, Lithium Battery 220V
$1,001.91CAD / Each
Adam Equipment SHS 300A Crane Scale, 150kg with 0.02kg Readability, Led, Lithium Battery 220V 1171040 Mfr # SHS 150
COMPARE
150 331.1 Crane Scale, 150kg with 0.02kg Readability, Led, Lithium Battery 220V
$1,038.74CAD / Each
Adam Equipment SHS 600A (220V) Crane Scale, 300kg with 0.05kg Readability, Led, Lithium Battery 220V 1171041 Mfr # SHS 300
COMPARE
300 662.3 Crane Scale, 300kg with 0.05kg Readability, Led, Lithium Battery 220V
$1,053.47CAD / Each