Adam Equipment GKa(M) Industrial Scale Indicator
Designed for versatility, the GK indicator has the features needed for a number of applications
  • Full tare range
  • Easily accessible internal rechargeable battery
  • Large backlit LCD display with capacity tracker
  • Battery operation with auto-off to save battery life
  • Parts counting with freely selectable sample sizes
  • Check weighing LED lights to show limits
  • Automatic accumulation
  • Bi-direcctional RS-232 output for connection to computers and printers
The GK offers an affordable indicator that is packed with smart features that will make weighing quick and easy. The built-in check weighing function allows the users to easily view if their samples are above, below or at set targets. With a full keypad, counting applications are achieved
with the same indicator, making the GK totally flexible for numerous uses.
The GK comes with a wall-mounted bracket for easy and secure mounting, with the indicator housing made of solid ABS plastic. The bright backlit LCD display features a capacity tracker to show users the total capacity that they have used to prevent potential overloads. The large digits make readings effortless.
Whether using the GK for basic weighing or taking advantage of the parts counting, check weighing or percentage weighing functions, these indicators offer a simple user interface for quick and hassle-free weight readings.
$574.62 - $744.06CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
Adam Equipment Gka Scale Indicator with Numeric Keypad and Checkweigh Lights 1171050 Mfr # GKA
COMPARE
100 to 240 Scale Indicator with Numeric Keypad and Checkweigh Lights
$574.62CAD / Each
Adam Equipment GKaM NTEP Scale Indicator with Numeric Keypad and Checkweigh Lights 1171052 Mfr # GKAM
COMPARE
100 to 240 NTEP Scale Indicator with Numeric Keypad and Checkweigh Lights
$744.06CAD / Each