Adam Equipment BCT Industrial Bench Counting Scales with Built-In Advanced Label Printer
Confidently count and weigh, then print labels in style
  • Print customized barcodes, QR codes, and custom logos on 50 mm wide labels
  • Durable stainless steel platform is removable and easy to clean
  • Store up to 1006 PLUs or piece weights
  • Color-coded keypad on indicator enhances efficiency
  • Available in a range of capacities and readabilities
This rugged bench counting scale features the AE 504 indicator with multi-line display and advanced printing capabilities, making it a first-class choice for your industrial counting and weighing tasks, while offering excellent value. Get dependably accurate results for parts counting, check counting, averaging, checkweighing, and more. The built-in integrated printer prints weights, bar codes, QR codes, pictures, and custom logos on 50 mm wide labels; supports multiple languages.
Large, backlit LCD makes it visible in any lighting condition and features a sealed color-coded numeric keypad which allows you quick and easy selection of over 1,000 PLUs or to manually enter tare weights, unit weights, sample sizes, and other frequently used functions. The backlight on the LCD can be set to "always on," "always off" or "lit only when weighing." Adjustable leveling feet optimize scale setup, RS-232 interface connects to computers/printers, and the included rechargeable battery allows you to use the scale almost anywhere. Programmable auto power-off saves energy.
$2,018.54 - $2,571.07CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (lbs) Readability (g)
Availability Pricing
Adam Equipment BCT 35A Industrial Bench Counting Scale with Built-In Advanced Label Printer, 35lb/6kg, 11.8" x 15.7" Platform 1113011 Mfr # BCT 35A
COMPARE
External calibration 35 0.5
$2,018.54CAD / Each
Adam Equipment BCT 65A Industrial Bench Counting Scale with Built-In Advanced Label Printer, 65lb/30kg, 11.8" x 15.7" Platform 1113012 Mfr # BCT 65A
COMPARE
External 65 1
$2,099.58CAD / Each
Adam Equipment BCT 130A Industrial Bench Counting Scale with Built-In Advanced Label Printer, 130lb/60kg, 11.8" x 15.7" Platform 1113013 Mfr # BCT 130A
COMPARE
External calibration 130 2
$2,099.58CAD / Each
Adam Equipment BCT165a Industrial Bench Counting Scale with Built-In Advanced Label Printer, 165lb/75kg, 11.8" x 15.7" Platform 1113014 Mfr # BCT 165A
COMPARE
External 165 5
$2,342.69CAD / Each
Adam Equipment BCT 330A Industrial Bench Counting Scale with Built-In Advanced Label Printer, 330lb/150kg, 15.7 " x 19.7"  Platform 1113015 Mfr # BCT 330A
COMPARE
External calibration 330 10
$2,342.69CAD / Each
Adam Equipment BCT660a Industrial Bench Counting Scale with Built-In Advanced Label Printer, 660lb/300kg, 15.7 " x 19.7"  Platform 1113016 Mfr # BCT 660A
COMPARE
External 660 20
$2,571.07CAD / Each