AccuBlock Digital Dry Bath Blocks
Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
  • PopStopper and AccuRack accessory sold seperately
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Thermometer wells available in all blocks except those used with plates.
$212.17 - $560.99CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mm) Description
Availability Pricing
Labnet D1101 Block, solid for machining, no holes 3788499 Mfr # D1101
COMPARE
0 Labnet AccuBlock D1101 Block, solid for machining, no holes
$212.17CAD / Each
Labnet D1102 Block, 0.2mL PCR tubes or 6 x 0.2mL strips 3788500 Mfr # D1102
COMPARE
48 Labnet AccuBlock D1102 Block, 48 x 0.2 mL PCR tubes or 6 x 0.2 mL strips
$250.11CAD / Each
Labnet D1102A Block, 20 x 2.0mL tubes 3788501 Mfr # D1102A
COMPARE
20 Labnet AccuBlock D1102A Block, 20 x 2.0 mL tubes
$246.42CAD / Each
Labnet D1105 Block, 24 x 0.5mL tubes 3788502 Mfr # D1105
COMPARE
24 Labnet AccuBlock D1105 Block, 24 x 0.5 mL tubes
$246.42CAD / Each
Labnet D1105A Bock, 24 x 1.5mL tubes 3788503 Mfr # D1105A
COMPARE
24 Labnet AccuBlock D1105A Block, 24 x 1.5 mL tubes
$288.05CAD / Each
Labnet D1110 Block, 20 x 10mm tubes 3788505 Mfr # D1110
COMPARE
20 10 Labnet AccuBlock D1110 Block, 20 x 10 mm tubes
$261.53CAD / Each
Labnet D1112 Block, 20 x 12mm tubes 3788506 Mfr # D1112
COMPARE
20 12 Labnet AccuBlock D1112 Block, 20 x 12 mm tubes
$236.48CAD / Each
Labnet D1113 Block, 20 x 13mm tubes 3788507 Mfr # D1113
COMPARE
20 13 Labnet AccuBlock D1113 Block, 20 x 13 mm tubes
$242.74CAD / Each
Labnet D1115-TALL Block, 12 x 15mL centrifuge tubes 3788508 Mfr # D1115-TALL
COMPARE
12 Labnet AccuBlock D1115-TALL Block, 12 x 15 mL centrifuge tubes
$321.94CAD / Each
Labnet D1116 Block, 12 x 16mm tubes 3788509 Mfr # D1116
COMPARE
12 16 Labnet AccuBlock D1116 Block, 12 x 16 mm tubes
$253.05CAD / Each
Labnet D1117 Block, 12 x 17mm tubes 3788510 Mfr # D1117
COMPARE
12 17 Labnet AccuBlock D1117 Block, 12 x 17 mm tubes
$227.64CAD / Each
Labnet D1120 Block, 6 x 20 mm tubes 3788511 Mfr # D1120
COMPARE
6 20 Labnet AccuBlock D1120 Block, 6 x 20 mm tubes
$232.80CAD / Each
Labnet D1125 Block, 6 x 25 mm tubes 3788512 Mfr # D1125
COMPARE
6 25 Labnet AccuBlock D1125 Block, 6 x 25 mm tubes
$261.53CAD / Each
Labnet D1150-TALL Block, 5 x 50mL centrifuge tubes 3788513 Mfr # D1150-TALL
COMPARE
5 Labnet AccuBlock D1150-TALL Block, 5 x 50 mL centrifuge tubes
$289.52CAD / Each
Labnet D1196-PCR Single Block 96 Well PCR Plate 3788514 Mfr # D1196-PCR
COMPARE
96 Labnet AccuBlock D1196-PCR Single Block 96 Well PCR Plate
$560.99CAD / Each
Showing 1- 15 of 15