A&D Weighing Phoenix GH Analytical Balances
A series of analytical balances which offer fast stabilization time and internal calibration.
  • Fast 2 second stabilization time
  • Resolution to 0.01mg
  • Anti-Static glass breeze break
  • Standard under hook
  • Data Memory up to 200 measurements
  • Standard RS-232C Output and WinCT software
A&D’s Phoenix GH-Series offers capacities from 120 grams to 250 grams with multiple weighing units, percentage and counting modes. Anti-Static glass helps to improve precision minimizing the effects of static electricity. Built memory allows the user to store up to 200 readings on the balance and output to a printer or computer via the RS-232C interface. All of the Phoenix GH-Series models come standard with a 5 Year Warranty.
$6,866.00 - $11,315.64CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
A&D Weighing GH-120 PHoenix Series Analytical Balance 120g x 0.1mgwith Internal Calibration 1111134 Mfr # GH-120
COMPARE
Internal 120 0.0001
$6,866.00CAD / Each
A&D Weighing GH-200 PHoenix Series Analytical Balance 220g x 0.1mg with Internal Calibration 1111135 Mfr # GH-200
COMPARE
Internal 200 0.0001
$7,772.13CAD / Each
A&D Weighing GH-300 PHoenix Series Analytical Balance 320g x 0.1mg with Internal Calibration 1111136 Mfr # GH-300
COMPARE
Internal 320 0.0001
$9,142.38CAD / Each
A&D Weighing GH-202 PHoenix Internal Cal Analytical Balance, 220/51g, 120V 1111137 Mfr # GH-202
COMPARE
Internal 220 0.0001
$10,284.26CAD / Each
A&D Weighing GH-252 PHoenix Internal Cal Analytical Balance, 250/101g x 0.1/0.01mg, 120V 1111138 Mfr # GH-252
COMPARE
Internal 250 0.0001
$11,315.64CAD / Each