A&D Weighing ION Analytical Balances
Weigh trace quantities with confidence – even fine powders
  • Built-in fan-less ionizer eliminates static interference
  • Interlocking doors allow you to open either door from opposite side
  • Auto-calibration when balance detects a charge in ambient temperature
  • Automatic response adjustment reduces effects from draft and vibration
  • Built-in sensors monitor, temperature, humidity, vibration and atmospheric pressure
  • Be compliant -- external data logger stores up to 5000 weighing results
These balances use a fan-less, integrated, direct current ionizer to neutralize charged samples. A green LED lights up when the ionizer is on. Transparent metal coating on the glass further minimizes the effects of static.
A bight backlit LCD allows you to easily read measurements. The units also feature one-touch digital internal calibration with auto self-cal (ASC), dual draft rings to reduce draft interferences, and a removable separation plate for weighing taller objects.
$8,287.82 - $9,945.38CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
A&D Weighing BM-200 Ion Analytical Balance, 220 g x 0.1 mg with Internal Calibration 1111092 Mfr # BM-200
COMPARE
Internal 220
$9,945.38CAD / Each
A&D Weighing BM-300 Ion Analytical Balance, 320 g x 0.1 mg with Internal Calibration 1111093 Mfr # BM-300
COMPARE
Internal 320 0.1
$8,287.82CAD / Each
A&D Weighing BM-500 Ion Analytical Balance, 520 g x 0.1 mg with Internal Calibration 1111094 Mfr # BM-500
COMPARE
Internal 520
$9,805.41CAD / Each