A&D Weighing Gemini GR Analytical Autocalibrating Balances
Automatic self-calibration adjusts for ambient temperature changes
  • Data memory function stores up to 200 sets of data
  • Balance ID for GLP applications outputs date, time, balance number, and calibration weight
  • Parts counting, percent weighing, below-balance weighing, and multiple weighing units
  • Rear-mount motor drive assembly and antistatic glass offer superior weighing accuracy
  • Splashproof keypad and display withstand messy lab environments
These balances are easy to use and calibrate and adjust their settings based on environmental changes. Balances feature a larger weighing chamber and smaller footprint than conventional analytical balances.
$5,797.79 - $9,319.19CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
A&D Weighing GR-120 Gemini Autocalibrating Analytical Balance, 120g x 0.1mg 1110060 Mfr # GR-120
COMPARE
Internal 120
$5,797.79CAD / Each
A&D Weighing GR-200 Gemini Autocalibrating Analytical Balance, 210g x 0.1mg 1110070 Mfr # GR-200
COMPARE
Internal 210
$6,578.69CAD / Each
A&D Weighing GR-202 Gemini Autocalibrating Analytical Balance, 42g/210g x 0.01/0.1mg 1110080 Mfr # GR-202
COMPARE
Internal 210
$9,319.19CAD / Each
A&D Weighing GR-300 Gemini Autocalibrating Analytical Balance 310g x 0.1mg 1111048 Mfr # GR-300
COMPARE
Internal 310
$8,192.05CAD / Each