A&D Weighing AD-4212A Production Weighing System with Display
High-precision weighing with a 0.8-second response time
  • FAST, MID, and SLOW stabilization settings let you choose the right combination of response time and sensitivity for your application
  • Compact footprint and mounting fixtures make integration into existing production line systems easy
  • Create your own weighing pan to better suit your samples
  • Separate sensor and display units are perfect for isolated weighing — such as inside vacuum chambers, fume hoods and glove boxes
  • Display unit can be panel or wall mounted
  • Patented shock absorber fitted under the weighing pan protects sensor from vertical and transverse overloads
  • Standard bidirectional RS-232 interface connects to PC for data download and control
  • Output conforms to GLP/GMP requirements
The extremely quick Super Hybrid Sensor technology makes this balance ideal for your high-speed weighing and filling, checkweighing, and remote weighing applications. Response times are as low as 0.5 seconds for weighing in a fast-paced environment or filling operations. The compact weighing sensor can be placed anywhere in a production line and allows for a custom-designed weighing pan. The AD-4212 series of weighing sensors provides GLP/GMP compliance with data output of weight and calibration data, as well as unit mass information for the counting mode or over/under limit values.
The bright vacuum fluorescent display included with the weighing sensor is IP54-rated for use in virtually any environment and can be located up to six feet from the weighing sensor. The data memory function provides storage of 200 weight measurements, 100 weight measurements with time and date, 50 calibration and calibration test measurements, 20 unit weight measurements for counting mode or 20 comparator settiing limits values.
The AD-4212A sensor for auto power on/off function and the lightweight yet durable painted aluminum housing.
$9,098.18 - $10,402.13CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing AD-4212A-100 Production System Display and Aluminum Housing, 110gx 0.1mg 1111202 Mfr # AD-4212A-100
COMPARE
110 115
$10,350.56CAD / Each
A&D Weighing AD-4212A-200 Production System Display and Aluminum Housing 210gx 1mg 1111203 Mfr # AD-4212A-200
COMPARE
210 115
$9,098.18CAD / Each
A&D Weighing AD-4212A-600 Production System Display and Aluminum Housing 610gx 1mg 1111204 Mfr # AD-4212A-600
COMPARE
610 115
$10,041.15CAD / Each
A&D Weighing AD-4212A-1000 Production System Display and Aluminum Housing, 1100Gx 0.001g 1111205 Mfr # AD-4212A-1000
COMPARE
1100 115 0.001
$10,402.13CAD / Each