A&D Phoenix Series Analytical Balances
Antistatic coating on glass minimizes static effects for more accurate weighing
  • Large weighing chamber allows weighing in large containers — up to 250 mL volumetric flask
  • Double spill protection around and below weighing pan protect vulnerable areas from spills
  • Diecast aluminum alloy housing offers durability and extended life
$6,866.00 - $11,315.64CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
A&D Weighing GH-120 PHoenix-Series Analytical Balance 120G x 0.1 Mg 1110050 Mfr # GH-120
COMPARE
Internal 120 0.0001
$6,866.00CAD / Each
A&D Weighing GH-200 PHoenix-Series Analytical Balance 220g x 0.1 Mg 1110052 Mfr # GH-200
COMPARE
Internal 220 0.0001
$7,772.13CAD / Each
A&D Weighing GH-300 PHoenix-Series Analytical Balance 320g x 0.1 Mg 1110054 Mfr # GH-300
COMPARE
Internal 320 0.0001
$9,142.38CAD / Each
A&D Weighing GH-202 PHoenix-Series Analytical Balance 50/220g x 0.01/0.1 Mg 1110056 Mfr # GH-202
COMPARE
Internal 220 1E-05
$10,284.26CAD / Each
A&D Weighing GH-252 PHoenix-Series Analytical Balance 100/250g x 0.01/0.1 Mg 1110058 Mfr # GH-252
COMPARE
Internal 250 0.0001
$11,315.64CAD / Each