A&D FZ-iWP Toploading Balances
A faster, more compact precision balance – with 1-second stabilization time
  • IP65 waterproof and dustproof
  • GLP, GMP, GCP, and ISO compliant printouts via USB interface
  • Easy-to-use digital calibration function with external calibration mass simplifies balance calibration procedures
  • Animal weighing, below-balance weighing, capacity indicator function, auto power on/off, and percent weighing
  • Internal calibration
A&D’s new FX-i series has the smallest footprint of most balances in its class. The 25% smaller footprint uses less bench space with all the capacities of larger toploading balances. The improved Super Hybrid Sensor (SHS) uses half the current of full-size toploaders, allowing the optional battery to last up to 8 hours on a single charge.
The FX-i offers standard RS-232 and the new Statistical Calculation Function (SCF) provides statistical data display or output of weighed samples. The balances feature 12 different weighing units and offer percentage and counting functions.
$3,366.70 - $3,779.25CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing FZ-120IWP Internal Cal Waterproof Toploading Balance, 122g x 0.001g, 115V 1111037 Mfr # FZ-120IWP
COMPARE
122 115 / 230 0.001
$3,366.70CAD / Each
A&D Weighing FZ-200IWP Internal Cal Waterproof Toploading Balance, 220g x 0.001g, 115V 1111038 Mfr # FZ-200IWP
COMPARE
220 115 / 230 0.001
$3,491.93CAD / Each
A&D Weighing FZ-300iWP Internal Cal Waterproof Toploading Balance, 320g x 0.001g, 115V 1111039 Mfr # FZ-300IWP
COMPARE
320 115 / 230 0.001
$3,771.88CAD / Each
A&D Weighing FZ-1200iWP Internal Cal Waterproof Toploading Balance, 1220g x 0.01g, 115V 1111041 Mfr # FZ-1200IWP
COMPARE
1220 115 / 230 0.01
$3,366.70CAD / Each
A&D Weighing FZ-2000iWP Internal Cal Waterproof Toploading Balance, 2200g x 0.01g, 115V 1111042 Mfr # FZ-2000IWP
COMPARE
2200 115 / 230 0.01
$3,491.93CAD / Each
A&D Weighing FZ-3000iWP Internal Cal Waterproof Toploading Balance, 3200g x 0.01g, 115V 1111043 Mfr # FZ-3000IWP
COMPARE
3200 115 / 230 0.01
$3,779.25CAD / Each