A&D FZ-i Toploading Balances
A faster, more compact precision balance – with 1-second stabilization time
  • GLP, GMP, GCP, and ISO compliant printouts via USB interface
  • Easy-to-use digital calibration function with external calibration mass simplifies balance calibration procedures
  • Animal weighing, below-balance weighing, capacity indicator function, auto power on/off, and percent weighing
  • Internal calibration
A&D’s new FX-i series has the smallest footprint of most balances in its class. The 25% smaller footprint uses less bench space with all the capacities of larger toploading balances. The improved Super Hybrid Sensor (SHS) uses half the current of full-size toploaders, allowing the optional battery to last up to 8 hours on a single charge.
The FX-i offers standard RS-232 and the new Statistical Calculation Function (SCF) provides statistical data display or output of weighed samples. The balances feature 12 different weighing units and offer percentage and counting functions.
$2,342.69 - $3,072.02CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing FZ-120i Internal Calibration Toploading Balance, 122g x 0.001g, 115V 1111031 Mfr # FZ-120I
COMPARE
122 115 / 230 0.001
$2,342.69CAD / Each
A&D Weighing FZ-200I Internal Calibration Toploading Balance, 220g x 0.001g, 115V 1111032 Mfr # FZ-200I
COMPARE
220 115 / 230 0.001
$2,467.93CAD / Each
A&D Weighing FZ-300I Internal Calibration Toploading Balance, 320g x 0.001g, 115V 1111033 Mfr # FZ-300I
COMPARE
320 115 / 230 0.001
$2,622.63CAD / Each
A&D Weighing FZ-1200I Internal Calibration Toploading Balance, 1220g x 0.01g, 115V 1111034 Mfr # FZ-1200I
COMPARE
1220 115 / 230 0.01
$2,519.50CAD / Each
A&D Weighing FZ-2000i Internal Calibration Toploading Balance, 2200g x 0.01g, 115V 1111035 Mfr # FZ-2000I
COMPARE
2200 115 / 230 0.01
$2,607.90CAD / Each
A&D Weighing FZ-3000i Internal Calibration Toploading Balance, 3200g x 0.01g, 115V 1111036 Mfr # FZ-3000I
COMPARE
3200 115 / 230 0.01
$2,688.94CAD / Each
A&D Weighing FZ-500i Toploading Balance, 520g x 0.001g Int.Calibration, Comparator, RS-232 1111097 Mfr # FZ-500I
COMPARE
520 115 / 230 0.001
$2,990.98CAD / Each
A&D Weighing FZ-5000I Toploading Balance, 5200g x 0.01g Int.Calibration, Comparator, RS-232 1111098 Mfr # FZ-5000I
COMPARE
5200 115 / 230 0.01
$3,072.02CAD / Each
Showing 1- 8 of 8