A&D FX-iWP Waterproof Toploading Balances
A faster, more compact precision balance – with 1-second stabilization time
  • IP65 waterproof and dustproof
  • Large vacuum fluorescent display for easy-to-read weighing results
  • GLP, GMP, GCP, and ISO compliant printouts via USB interface
  • Easy-to-use digital calibration function with external calibration mass simplifies balance calibration procedures
  • Animal weighing, below-balance weighing, capacity indicator function, auto power on/off, and percent weighing
  • External calibration
A&D’s new FX-iWP series has the smallest footprint of most balances in its class. The 25% smaller footprint uses less bench space with all the capacities of larger toploading balances. The improved Super Hybrid Sensor (SHS) uses half the current of full-size toploaders, allowing the optional battery to last up to 8 hours on a single charge.
The FX-iWP offers standard RS-232 and the new Statistical Calculation Function (SCF) provides statistical data display or output of weighed samples. The balances feature 12 different weighing units and offer percentage and counting functions.
$2,725.77 - $3,079.39CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing FX-120IWP Toploading Balance, 122g x 0.001g Ext.Calibration, Comparator, RS-232 1111270 Mfr # FX-120IWP
COMPARE
122 115 / 230 0.001
$2,740.51CAD / Each
A&D Weighing FX-200IWP Toploading Balance, 220g x 0.001g Ext.Calibration, Comparator, RS-232 1111271 Mfr # FX-200IWP
COMPARE
220 115 / 230 0.001
$2,880.48CAD / Each
A&D Weighing FX-300IWP Toploading Balance, 320g x 0.001g Ext.Calibration, Comparator, RS-232 1111272 Mfr # FX-300IWP
COMPARE
320 115 / 230 0.001
$3,079.39CAD / Each
A&D Weighing FX-1200IWP Toploading Balance, 1220g x 0.01g Ext.Calibration, Comparator, RS-232 1111273 Mfr # FX-1200IWP
COMPARE
1220 115 / 230 0.01
$2,725.77CAD / Each
A&D Weighing FX-2000IWP Toploading Balance, 2200g x 0.01g Ext.Calibration, Comparator, RS-232 1111274 Mfr # FX-2000IWP
COMPARE
2200 115 / 230 0.01
$2,880.48CAD / Each
A&D Weighing FX-3000IWP Toploading Balance, 3200g x 0.01g Ext.Calibration, Comparator, RS-232 1111275 Mfr # FX-3000IWP
COMPARE
3200 115 / 230 0.01
$3,072.02CAD / Each