A&D FX-i NTEP Approved Toploading Balances
A faster, more compact precision balance – with 1-second stabilization time
  • NTEP Approved
  • GLP, GMP, GCP, and ISO compliant printouts via USB interface
  • Easy-to-use digital calibration function with external calibration mass simplifies balance calibration procedures
  • Animal weighing, below-balance weighing, capacity indicator function, and percent weighing
  • Features a capacity indicator and below balance weighing hook
  • External calibration
A&D’s FX-i series has the smallest footprint of most balances in its class. The 25% smaller footprint uses less bench space with all the capacities of larger toploading balances. The improved Super Hybrid Sensor (SHS) uses half the current of full-size toploaders, allowing the optional battery to last up to 8 hours on a single charge.
The FX-i offers standard RS-232 and the new Statistical Calculation Function (SCF) provides statistical data display or output of weighed samples. The balances feature 12 different weighing units and offer percentage and counting functions.
$2,202.72 - $2,467.93CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing FX-120IN NTEP Toploading Balance 122g x 0.001g 1111581 Mfr # FX-120IN
COMPARE
122 115 0.001
$2,202.72CAD / Each
A&D Weighing FX-2000iN NTEP Toploading Balance 2200g x 0.01g 1111582 Mfr # FX-2000IN
COMPARE
2200 115 0.01
$2,254.29CAD / Each
A&D Weighing FX-200iN NTEP Toploading Balance 220g x 0.001g 1111583 Mfr # FX-200IN
COMPARE
220 115 0.001
$2,224.82CAD / Each
A&D Weighing FX-3000IN NTEP Toploading Balance 3200g x 0.01g 1111584 Mfr # FX-3000IN
COMPARE
3200 115 0.01
$2,467.93CAD / Each
A&D Weighing FX-300iN NTEP Toploading Balance 320g x 0.001g 1111585 Mfr # FX-300IN*
COMPARE
320 115 0.001
$2,202.72CAD / Each