Zefon MoldSNAP™ Cassette, 50/BX
Representative image only.
Zefon MoldSNAP™ Cassette, 50/BX
Zefon | Item: 6003091
A cost effective first-line mold screening tool that efficiently and reliably collects mold and allows for simple lab analysis
$366.14CAD / Box of 50
$366.14CAD / Box of 50
Product Description
Key Features
  • 5 Liters x 5 minute sampling
  • Industry proven collection and analysis method
  • Quick, low volume sample
  • Easy for labs to analyze
  • Pre-loaded so no media to prep in the field
  • 1 year warranty
More About this Item
MoldSNAP™ is designed as a first-line mold screening tool that can be easily used by home inspectors to get preliminary information on whether a mold problem may exist and further investigation is warranted. MoldSNAP is a type of "spore trap" impactor that uses slit impaction as the collection mechanism. This is important as slit impaction is not only the most reliable collection method, it is an industry-recognized method that allows superior mold spore collection.
Requires the IAQ sampling pump 60062-07 or 60062-08. Not compatible with the Zefon Bio-Pump Plus.
1 Year Warranty
Specifications
  • Parameters MeasuredMold sampling
  • DescriptionMoldSNAP™ Cassette, 50/BX