YSI Thermoreactor, 24 Vials, 115 V

Representative image only.

YSI Thermoreactor, 24 Vials, 115 V

YSI | Mfr: 1P22-2Y| Item: 9955025

US EPA-approved COD test method

$2,291.12CAD / Each

$2,291.12CAD / Each
Product Description

Key Features

  • Premeasured COD reagents cover three test ranges
  • 430 nm single-wavelength colorimeter is specific to COD testing
  • Digest up to 24 samples simultaneously with thermal reactor
  • 2 year warranty

More About this Item

Testing requires the purchase of all three components: a COD colorimeter, a thermal reactor, and COD test reagents.

Thermal Reactor for COD digests up to 24 samples simultaneously. Works with the YSI test kits or on any 16-mm diameter COD vial. Features fully selectable temperature range from 25 to 170°C; with eight fixed and eight user-selectable programs.

Two year warrantyConformite Europeenne
Specifications
  • TestChemical oxygen demand (COD)
  • DescriptionThermoreactor, 24 Vials, 115 V
  • Warranty2 year
Reviews
Q&A