YSI DSS Pro Instrument Sensor, Turbidity (NePHelometer)
Representative image only.
YSI DSS Pro Instrument Sensor, Turbidity (NePHelometer)
YSI | Mfr: 626901| Item: 5935412
Choose the sensors necessary for the application
$2,056.85CAD / Each
$2,056.85CAD / Each
Buy It Together
Current Item
YSI DSS Pro Instrument Sensor, Turbidity (NePHelometer)
YSI DSS Pro Instrument Sensor, Turbidity (NePHelometer)
5935412
$2,056.85CAD / Each
+
YSI 605101 Pro Series Replacement pH Electrode
5935220
$346.25CAD / Each
+
YSI ProDSS Instrument Cable, 1 M
5935420
$3,527.30CAD / Each
+
YSI 626870-1 ProDSS Multiparameter Meter without GPS
5935400
$3,340.18CAD / Each
=
Total Price 9270.58 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Digital smart sensor stores calibration data — change sensors without recalibrating
  • 1 year warranty
More About this Item
The ProDSS sensor housing accepts four user-replaceable sensors at a time and the universal connector system allows any sensor to connect with any housing port.
Turbidity sensor (59354-12) is an optical nephelometric type with a 90° scatter.
1 Year Warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • Min Turbidity0 to 4000 FNU
  • Turbidity Resolution0.1 FNU
  • Turbidity Accuracy±0.3 FNU or ±2% of rdg, whichever is greater (up to 999 FNU); ±5% of rdg (above 999 FNU)
  • Data Logging (points)Calibration data
  • DescriptionPro Instrument Sensor, Turbidity (NePHelometer)