YSI 626906 ProDSS Instrument Sensor with Module, Ammonium
Representative image only.
YSI 626906 ProDSS Instrument Sensor with Module, Ammonium
YSI | Mfr: 626906| Item: 5935415
Choose the sensors necessary for the application
$1,066.73CAD / Each
$1,066.73CAD / Each
Buy It Together
Current Item
YSI 626906 ProDSS Instrument Sensor with Module, Ammonium
YSI 626906 ProDSS Instrument Sensor with Module, Ammonium
5935415
$1,066.73CAD / Each
+
YSI 605101 Pro Series Replacement pH Electrode
5935220
$346.25CAD / Each
+
YSI ProDSS Instrument Cable, 1 M
5935420
$3,527.30CAD / Each
+
YSI 626870-1 ProDSS Multiparameter Meter without GPS
5935400
$3,340.18CAD / Each
=
Total Price 8280.46 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Digital smart sensor stores calibration data — change sensors without recalibrating
  • 1 year warranty
More About this Item
The ProDSS sensor housing accepts four user-replaceable sensors at a time and the universal connector system allows any sensor to connect with any housing port.
Ion selective electrodes (59354-15, 59354-16, and 59354-17) have replaceable sensor modules.
1 Year Warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • Min ISE0 to 200 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±10% of rdg or 2 ppm, whichever is greater
  • Data Logging (points)Calibration data
  • DescriptionProDSS Instrument Sensor with Module, Ammonium