YSI 626903 ProDSS Instrument Sensor, pH Electrode
Representative image only.
YSI 626903 ProDSS Instrument Sensor, pH Electrode
YSI | Mfr: 626903| Item: 5935413
Choose the sensors necessary for the application
$839.83CAD / Each
$839.83CAD / Each
Buy It Together
Current Item
YSI 626903 ProDSS Instrument Sensor, PH Electrode
YSI 626903 ProDSS Instrument Sensor, PH Electrode
5935413
$839.83CAD / Each
+
YSI 605101 Pro Series Replacement pH Electrode
5935220
$346.25CAD / Each
+
YSI 626870-1 ProDSS Multiparameter Meter without GPS
5935400
$3,340.18CAD / Each
+
YSI ProDSS Instrument Cable, 1 M
5935420
$3,527.30CAD / Each
=
Total Price 8053.56 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Digital smart sensor stores calibration data — change sensors without recalibrating
  • 1 year warranty
More About this Item
The ProDSS sensor housing accepts four user-replaceable sensors at a time and the universal connector system allows any sensor to connect with any housing port.
pH sensor (59354-13) is a combination type with an Ag/AgCl gel-filled glass bulb. Sensor module is replaceable.
1 Year Warranty
Specifications
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01
  • pH Accuracy±0.2
  • Data Logging (points)Calibration data
  • DescriptionProDSS Instrument Sensor, pH Electrode