YSI 626902 ProDSS Instrument Sensor, Conductivity and Temperature
Representative image only.
YSI 626902 ProDSS Instrument Sensor, Conductivity and Temperature
YSI | Mfr: 626902| Item: 5935411
Choose the sensors necessary for the application
$1,315.74CAD / Each
$1,315.74CAD / Each
Buy It Together
Current Item
YSI 626902 ProDSS Instrument Sensor, Conductivity and Temperature
YSI 626902 ProDSS Instrument Sensor, Conductivity and Temperature
5935411
$1,315.74CAD / Each
+
YSI 605101 Pro Series Replacement pH Electrode
5935220
$346.25CAD / Each
+
YSI ProDSS Instrument Cable, 1 M
5935420
$3,527.30CAD / Each
+
YSI 626870-1 ProDSS Multiparameter Meter without GPS
5935400
$3,340.18CAD / Each
=
Total Price 8529.47 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Digital smart sensor stores calibration data — change sensors without recalibrating
  • 1 year warranty
More About this Item
The ProDSS sensor housing accepts four user-replaceable sensors at a time and the universal connector system allows any sensor to connect with any housing port.
Conductivity cell (59354-11) has four electrodes and houses the Automatic Temperature Compensation (ATC) thermistor.
1 Year Warranty
Specifications
  • Min Conductivity0 to 200 mS
  • Conductivity Accuracy±0.5% of reading or 0.001 mS/cm, whichever is greater (up to 100 mS/cm); ±1.0% of reading from 100 to 200 mS/cm
  • Min TDS0 to 100 ppt
  • Data Logging (points)Calibration data
  • DescriptionProDSS Instrument Sensor, Conductivity and Temperature