YSI 626900 ProDSS Instrument Sensor, Optical Dissolved Oxygen (OD O)
Representative image only.
YSI 626900 ProDSS Instrument Sensor, Optical Dissolved Oxygen (OD O)
YSI | Mfr: 626900| Item: 5935410
Choose the sensors necessary for the application
$1,869.73CAD / Each
$1,869.73CAD / Each
Buy It Together
Current Item
YSI 626900 ProDSS Instrument Sensor, Optical Dissolved Oxygen (OD O)
YSI 626900 ProDSS Instrument Sensor, Optical Dissolved Oxygen (OD O)
5935410
$1,869.73CAD / Each
+
YSI 605101 Pro Series Replacement pH Electrode
5935220
$346.25CAD / Each
+
YSI ProDSS Instrument Cable, 1 M
5935420
$3,527.30CAD / Each
+
YSI 626870-1 ProDSS Multiparameter Meter without GPS
5935400
$3,340.18CAD / Each
=
Total Price 9083.46 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Digital smart sensor stores calibration data — change sensors without recalibrating
  • 1 year warranty
More About this Item
The ProDSS sensor housing accepts four user-replaceable sensors at a time and the universal connector system allows any sensor to connect with any housing port.
Dissolved oxygen sensor (59354-10) features optical technology utilizing lifetime-based detectors.
1 Year Warranty
Specifications
  • Min Dissolved Oxygen0 to 500%, 0 to 50 ppm
  • Dissolved Oxygen Resolution0.1 or 0.01 ppm; 0.1 or 1% saturation
  • Dissolved Oxygen Accuracy±1% of rdg or 1% saturation, whichever is greater (up to 200%), ±8% of rdg (above 200%); ±0.1 ppm or 1% of rdg, whichever is greater (up to 20 ppm), ±8% of rdg (above 20 ppm)
  • Data Logging (points)Calibration data
  • DescriptionProDSS Instrument Sensor, Optical Dissolved Oxygen (OD O)