Wildco 1750-G30 Petersen grab dredge, 12" x 12" sampling area
Representative image only.
Wildco 1750-G30 Petersen grab dredge, 12" x 12" sampling area
Wildco | Mfr: 1750-G30| Item: 0547200
Quickly sample sediment from various bodies of water
$6,406.37CAD / Each
$6,406.37CAD / Each
Product Description
Key Features
  • Dredge is made of zinc-plated steel
  • Includes jaw weights and a safety lock to hold jaws open during handling and transporting
  • Sampling area is 12" x 12". Recommend using a winch and aircraft cable to operate this dredge
  • 3 month warranty
More About this Item
Sample sand, marl, gravel, clay, and hard-bottom surfaces of deep lakes and oceans. Dredge provides deep-biting penetration with its tapered jaws and 70-lb weight. Vent holes in the jaws reduce frontal shock waves. A latch-type mechanism closes jaws when dredge hits bottom.
3 Month Warranty
Specifications
  • MaterialZinc-plated Steel
  • DescriptionPetersen grab dredge, 12" x 12" sampling area