Whatman 6705-3602 CYTIVA Whatman™ Polycap AS NYL Capsule Filter, 0.2um, 400cm2, 1/4-3/8" ID; 1/Pk
Representative image only.
Whatman 6705-3602 CYTIVA Whatman™ Polycap AS NYL Capsule Filter, 0.2um, 400cm2, 1/4-3/8" ID; 1/Pk
Whatman | Mfr: 6705-3602| Item: 2970040
Nylon membranes are hydrophilic and low in extractables
$286.65CAD / Each
$286.65CAD / Each
Product Description
Key Features
  • Nylon filters include a prefilter to prolong life of the membrane
  • Use for filtering aqueous solutions
More About this Item
Capsules feature sturdy PP housings with fused seals—no glues or adhesives to contaminate filtrate. Capsules include a luer lock vent to guarantee that no filtration area is wasted. Use vent to draw solution into the capsule or let air escape from the capsule or sample port.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
Specifications
  • Membrane MaterialNylon
  • Pore Size (µm)0.2
  • Effective Filter Area (cm²)440
  • Connections1/4" to 3/8" stepped barb
  • DescriptionPolycap AS NYL Capsule Filter, 0.2um, 400cm2, 1/4-3/8" ID; 1/Pk