Whatman 6704-7500 CYTIVA Whatman™ In-Line Carbon Filter Capsule, 96 x 66 mm

Representative image only.

Whatman 6704-7500 CYTIVA Whatman™ In-Line Carbon Filter Capsule, 96 x 66 mm

Whatman | Mfr: 6704-7500| Item: 2970810

Disposable filter capsule contains high purity activated carbon and glass microfiber HEPA filter

$446.00CAD / Each

$446.00CAD / Each
Product Description

Key Features

  • Large surface area of activated carbon for effective operation
  • Two sizes of capsule available to suit your specific application
  • 45 day warranty

More About this Item

This filter is ideal for applications involving compressed air lines, vacuum pumps, or instrument exhaust emissions. The capsule contains a high-purity granular activated carbon and has a pleated glass microfiber HEPA filter at the base of the unit which retains 99.97% of all particles greater than 0.3µm. The in-line filter device features a high purity polypropylene housing with a 1/2" (12 mm) hose barb inlet and 3/8" to 1/2" (10 to 12 mm) stepped barb outlet.

Typical applications include water, chemical and reagent purification, odor control, vacuum pump outlets, decolorizing and oil clarification. Maximum operating pressure is 60 psi (4.1 bar).

CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.

Specifications
  • Membrane MaterialActivated carbon with a pleated HEPA cartridge
  • Filter Size96 x 66
  • Housing MaterialPolypropylene
  • Connections1/2" hose barb by 3/8" to 1/2" hose barb
  • Surface Area (m²)26000
  • Particle Retention (µm)0.3
  • Particle Retention (%)99.97
  • Typical Water Flow Rate8.8LPM at 5 psi
  • Air Flow Rate0.3 psi: 72 SLPM; 1.6 psi: 212 SLPM
  • Max Pressure (PSI)60
  • Max Pressure4.1
  • DescriptionIn-Line Carbon Filter Capsule, 96 x 66 mm
  • Warranty45 days
Reviews
Q&A