Wearwell 24/Seven® Modular Industrial Anti-Fatigue Mat, Grease Resistant, Drainage, Black, 3 ft x 3 ft

Representative image only.

Wearwell 24/Seven® Modular Industrial Anti-Fatigue Mat, Grease Resistant, Drainage, Black, 3 ft x 3 ft

Wearwell | Mfr: 572.58X3X3GRBK| Item: 8185021

Modular anti-fatigue, anti-slip mats keep employees safe and comfortable

$155.81CAD / Each

$155.81CAD / Each
Product Description

Key Features

  • Keep workers productive, safe, and comfortable all day, every day
  • Extremely versatile — mats interlock on all sides to cover entire floor areas, long assembly lines, or stand alone as single workstation mats
  • Tough enough to handle 24/7 shift operations
  • For extra safety, attach optional 2" x 39" yellow safety edges
  • Ideal for use with water or debris
  • Drainage style mates with perforated 15/16" hold maximizes drainage in wet workspaces
  • 1 year warranty

More About this Item

The 24/Seven® modular mat provides enhanced comfort and safety with anti-fatigue and cushioned support. Customize matting to your desired size using these 3 ft x 3 ft interlocking modules. The hidden interlocking system is designed to connect on all four sides to quickly cover large areas. Yellow beveled edging provides protection from tripping and a clean finished look; order separately.

Choose from three different rubber compounds based on environmental exposures.
Grease resistant model GR mats are suitable for water or dry debris.
Cutting-fluid resistant model CFR mats are ideal for mineral oil-based fluids.
Nitrile rubber model NBR mats stand up to petroleum-based fluids.

GRITSHIELD™ provides added traction where slip hazards are present. Solid design mats are for dry applications and open drainage models have large holes to allow fluid and debris to flow through.

1 Year Warranty
Specifications
  • MaterialRubber
  • Thickness (in)0.625
  • Thickness (mm)16
  • Width (in)36
  • Length (in)36
  • Width (cm)91
  • Length (cm)91
  • ColorBlack
  • DescriptionModular Industrial Anti-Fatigue Mat, Grease Resistant, Drainage, Black, 3 ft x 3 ft
  • Warranty1 year
Reviews
Q&A