Waring LB10G variable-speed blender, 1-liter, glass 120v
Representative image only.
Waring LB10G variable-speed blender, 1-liter, glass 120v
Waring | Mfr: LB10G| Item: 0424560
$3,027.82CAD / Each
$3,027.82CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Variable-speed blenders provide smooth, adjustable speed control from 1,000 to 22,000 rpm. Easily adjust at any speed for mixing, stirring, blending, or homogenizing applications. Both models include a container, lid, SS blades, and a 6-ft cord with U.S. standard plug. UL-listed.
UL Listed.1 Year Warranty
Specifications
  • Capacity (L)1
  • Min RPM1000
  • Max RPM22000
  • Motor TypeStandard
  • Power (VAC)120
  • Power (amps)4
  • Power (Hz)60
  • Container MaterialGlass
  • Descriptionvariable-speed blender, 1-liter, glass 120v