VistaLab Reagent Reservoir, Polystyrene, Sterile, 50 mL; 100/Cs

Representative image only.

VistaLab Reagent Reservoir, Polystyrene, Sterile, 50 mL; 100/Cs

VistaLab | Mfr: 3054-1006| Item: 0619872

Deep-well design allows you to pipette even small amounts of reagent

$175.33CAD / Case of 100

$175.33CAD / Case of 100
Product Description

Key Features

  • Trough-within-a-trough design to recover the “last few drops”
  • Pour-off spouts at every corner
  • Constructed of pyrogen-free polystyrene
  • 1 year warranty

More About this Item

These recyclable reagent reservoirs are available in a variety of volumes and packaging configurations. Constructed of pyrogen-free polystyrene, they will preserve the integrity of your sample. Well-marked graduated increments on the inside wall (all sizes except 100 mL) help you visualize reagent levels and minimize waste.

Wide base and sturdy construction provide stability to avoid spills. Unique trough design allows even small amounts of liquid to be drawn through all pipette tips. Trimmed base corners and distinct pour-off spouts make it easy to empty the reservoir without spills.

Order the reusable, nontoxic gel cooler to preserve the integrity of reagents and extend working time with full reservoir cooling. Custom fit for the 100-mL reagent reservoirs, the cooler will maintain your reagent at <41°F (5°C) for up to two hours.

1 Year Warranty
Specifications
  • SterileYes
  • Max Volume (mL)50
  • Number Of Channels1
  • Wetted MatterialPolystyrene
  • Package OfBulk Packed 100/Cs
  • DescriptionReagent Reservoir, Polystyrene, Sterile, 50 mL; 100/Cs
Reviews
Q&A