VF Workwear KEL2LB-ARGE Flame-Resistant Lab Coat, Light Blue, extra large
Representative image only.
VF Workwear KEL2LB-ARGE Flame-Resistant Lab Coat, Light Blue, extra large
VF Workwear | Mfr: KEL2LB-XLARGE (PACK BY PERSON)| Item: 8646833
Flame-resistant lab coats protect you when working with flammable chemicals
$198.54CAD / Each
BUY MORE & SAVE
  • 1-5$198.54 CAD / Each
  • 6+$178.69 CAD / Each
$198.54CAD / Each
BUY MORE & SAVE
  • 1-5$198.54 CAD / Each
  • 6+$178.69 CAD / Each
Product Description
More About this Item
Constructed of 100% cotton, lab coats feature three outside pockets, side vent openings, hemmed sleeve ends, and a lapel collar. Rated to Arc Thermal Protective Value (ATPV) of 7.7 calories/cm2 and a Hazard Risk Category (HRC) of HRC1.
Specifications
  • Length (in)41.5
  • Length (cm)105.41
  • MaterialCotton
  • SizeX-Large
  • ColorBlue
  • DescriptionFlame-Resistant Lab Coat, Light Blue, extra large