Velp Vreceiver TKN formula for 1 L solution, 40 g

Representative image only.

Velp Vreceiver TKN formula for 1 L solution, 40 g

Velp | Mfr: A00000411 | Item: 8658069

Expand the capabilities of your digestion system

$16.28CAD / Each

$16.28CAD / Each
Product Description

Key Features

  • Racks, caps, supports, stands, and more!
  • 2 year warranty

More About this Item

These accessories are compatible with Velp's DK and DKL Digestion Systerms.

Two year warranty
Specifications
  • DescriptionVreceiver TKN formula for 1 L solution, 40 g
  • Warranty2 year
Reviews
Q&A