Vaisala 6221 Plastic Grid Filter for Use with Hm46 Thermohygrometer Probe
Representative image only.
Vaisala 6221 Plastic Grid Filter for Use with Hm46 Thermohygrometer Probe
Vaisala | Mfr: 6221| Item: 3780122
$49.21CAD / Each
$49.21CAD / Each
Buy It Together
Current Item
Vaisala 6221 Plastic Grid Filter for Use with Hm46 Thermohygrometer Probe
Vaisala 6221 Plastic Grid Filter for Use with Hm46 Thermohygrometer Probe
3780122
$49.21CAD / Each
+ =
Total Price 1395.89 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Convenient parts and accessories for your thermohygrometer. Change capabilities, or keep a spare!
Two year warranty
Specifications
  • Min RH Range (%)0
  • Max RH Range (%)100
  • RH Accuracy±1% RH; ±2% RH above 90% RH
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • Temperature Accuracy±0.2°C
  • DescriptionPlastic Grid Filter for Use with Hm46 Thermohygrometer Probe