Uniweld HC5SMB Refrigerant Charging Hose, 5-ft length
Representative image only.
Uniweld HC5SMB Refrigerant Charging Hose, 5-ft length
Uniweld | Mfr: HC5SMB| Item: 1011260
Rugged nylon barrier hose for CFC, HCFC, and HFC refrigerants
$153.97CAD / Each
$153.97CAD / Each
Product Description
Key Features
  • Maximum working pressure is 800 psi; burst pressure of 4000 psi
  • 1/4" SAE fittings with 45° bend on one end
  • Each hose set is 5-ft in length and includes one red, yellow, and blue hose
  • Available with and without anti-blowback fittings
  • 2 year warranty
Two year warranty
Specifications
  • Length (in)60
  • DescriptionRefrigerant Charging Hose, 5-ft length