United Electric 4W3A01-T-H1 Temperature Switch, -50/1000F; 115V/240V/10A, 6' Mi

Representative image only.

United Electric 4W3A01-T-H1 Temperature Switch, -50/1000F; 115V/240V/10A, 6' Mi

United Electric | Mfr: 1XSWHHTH1| Item: 3900753

$2,453.19CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$2,453.19CAD / Each

Buy It Together

Current Item

United Electric 4W3A01-T-H1 Temperature Switch, -50/1000F; 115V/240V/10A, 6' Mi
United Electric 4W3A01-T-H1 Temperature Switch, -50/1000F; 115V/240V/10A, 6' Mi
3900753

$2,453.19CAD / Each
+ =
Total Price 2938.67 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 3 year warranty

More About this Item

The Preferred Choice for Plant Instrumentation Upgrades

  • Legendary UE Reliability
  • Hazardous Location
  • Class I, Div 2; Class II, Div. 2; Class III
  • Worldwide approvals and certifications
  • Large process display
  • 100% programmable set-point and deadband
  • Terminal block wiring
  • NEMA 4X / IP66 Die cast aluminum enclosure, epoxy powder coated, & gasketed

3 Year Warranty
Specifications
  • Min Temperature (° F)-50
  • Max Temperature (° F)1000
  • DescriptionTemperature Switch, -50/1000F; 115V/240V/10A, 6' Mi
  • Warranty3 year
Reviews
Q&A