United Electric 2W3A00-T-R1 Temperature Switch, -50/450F; 115V/100 Ma, 6' PTFE

Representative image only.

United Electric 2W3A00-T-R1 Temperature Switch, -50/450F; 115V/100 Ma, 6' PTFE

United Electric | Mfr: 1XSWHLTR1| Item: 3900751

$2,180.62CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$2,180.62CAD / Each

Buy It Together

Current Item

United Electric 2W3A00-T-R1 Temperature Switch, -50/450F; 115V/100 Ma, 6' PTFE
United Electric 2W3A00-T-R1 Temperature Switch, -50/450F; 115V/100 Ma, 6' PTFE
3900751

$2,180.62CAD / Each
+ =
Total Price 2666.10 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 3 year warranty

More About this Item

The Preferred Choice for Plant Instrumentation Upgrades

  • Legendary UE Reliability
  • Hazardous Location
  • Class I, Div 2; Class II, Div. 2; Class III
  • Worldwide approvals and certifications
  • Large process display
  • 100% programmable set-point and deadband
  • Terminal block wiring
  • NEMA 4X / IP66 Die cast aluminum enclosure, epoxy powder coated, & gasketed

3 Year Warranty
Specifications
  • Min Temperature (° F)-50
  • Max Temperature (° F)450
  • DescriptionTemperature Switch, -50/450F; 115V/100 Ma, 6' PTFE
  • Warranty3 year
Reviews
Q&A