Trajan SGE 005668 Gas Tight Syringe, Compatible with Agilent Autosampler, Fixed Needle, 100uL, 23-26 G; 1/Ea
Representative image only.
Trajan SGE 005668 Gas Tight Syringe, Compatible with Agilent Autosampler, Fixed Needle, 100uL, 23-26 G; 1/Ea
Trajan | Mfr: 005668| Item: 1600091
Designed and tested to meet critical autosampler specifications
$266.32CAD / Each
$266.32CAD / Each
Product Description
Key Features
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
  • 6 month warranty
More About this Item
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • Syringe TypeAutosampler
  • Syringe Capacity (µL)100
  • Needle IncludedY
  • Needle Tip StyleCone
  • Needle Gauge23/26
  • DescriptionTrajan SGE Gas Tight Syringe, Compatible with Agilent Autosampler, Fixed Needle, 100uL, 23-26 G; 1/Ea