Traceable Digital Pocket Thermometer Ultra; ±0.4°C Accuracy at Tested Points

Representative image only.

Traceable Digital Pocket Thermometer Ultra; ±0.4°C Accuracy at Tested Points

Traceable | Mfr: 4344TR| Item: 9876734

Get an updated reading every 10 seconds

$129.36CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$129.36CAD / Each

Buy It Together

Current Item

Traceable Digital Pocket Thermometer Ultra; ±0.4°C Accuracy at Tested Points
Traceable Digital Pocket Thermometer Ultra; ±0.4°C Accuracy at Tested Points
9876734

$129.36CAD / Each
+ =
Total Price 156.47 ADD 2 ITEMS TO CART
Product Description

Includes

Traceable® Certificate, protective case with clip, and battery.

Key Features

  • Wide temperature range from -50 to 300°C (-58 to 572°F)
  • Large, bright LCD with 3/8"H digits
  • 4-3/4"L x 0.19" dia stainless steel stem fits nicely into included protective sleeve
  • °C/°F selectable
  • ±0.4°C accuracy at tested points for high-precision monitoring
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

Easily take temperature measurements of soil, food products, and reagents. The 4-3/4" long piercing stainless steel probe easily handles liquids and semi-solids. Thermometers feature a wide temperature range of -50 to 300°C (-58 to 572°F) with readings updating every 10 seconds. A protective case with a clip makes it easy to carry around.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)572
  • Min Temperature (° C)-50
  • Max Temperature (° C)300
  • Accuracy±0.4°C at tested points; otherwise ±1°C
  • Resolution0.1° (–20 to 200°); otherwise 1°
  • Display Type3/8" LCD
  • Diameter (in)1 1/2
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.19
  • Probe Length (in)4 3/4
  • Battery1 1.5V Silver Oxide (included)
  • DescriptionDigital Pocket Thermometer Ultra; ±0.4°C Accuracy at Tested Points
  • Warranty1 Year
Reviews
Q&A