ThermoSafe 440 Mini Multipurpose Mailers, 24/cs

Representative image only.

ThermoSafe 440 Mini Multipurpose Mailers, 24/cs

ThermoSafe | Mfr: 440| Item: 0374120

Ship non-hazardous specimens safely

$249.00CAD / Case of 24

$249.00CAD / Case of 24

Buy It Together

Current Item

ThermoSafe 440 Mini Multipurpose Mailers, 24/cs
ThermoSafe 440 Mini Multipurpose Mailers, 24/cs
0374120

$249.00CAD / Case of 24
+
ThermoSafe 365 Shipping Container/Carton for 03727-01
0372723

$294.31CAD / Case of 300
+ =
Total Price 677.83 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

More About this Item

Protect your fragile samples with this expanded polystyrene shipper. Light weight minimizes postage costs. Strong white paperboard shipping carton (sold separately) has sure-lock tabs that eliminate taping or tying. Choose from tube mailers, slide mailer, mini mailer, multipurpose mailer, or aluminum shipping can.

Accomodates tubes, vials, ampules, small bottles, and tissue flasks with space for refrigerants.

Specifications
  • Inside Depth (in)3
  • Inside Length (in)5 7/8
  • Inside Width (in)4 3/8
  • Outside Width (in)6 1/2
  • Outside Length (in)8
  • Outside Depth (in)5 1/8
  • Inside Depth (cm)7.62
  • Inside Length (cm)14.9
  • Outside Width (cm)16.5
  • Outside Length (cm)20.3
  • Outside Depth (cm)13
  • Shipping Weight (oz)94
  • Shipping Weight (g)2676
  • DescriptionMini Multipurpose Mailers, 24/cs
Reviews
Q&A