ThermoSafe 369 Microscope Slide Mailer, holds 10 slides, 300/cs

Representative image only.

ThermoSafe 369 Microscope Slide Mailer, holds 10 slides, 300/cs

ThermoSafe | Mfr: 369| Item: 0375010

Ship non-hazardous specimens safely

$428.39CAD / Case of 300

$428.39CAD / Case of 300

Buy It Together

Current Item

ThermoSafe 369 Microscope Slide Mailer, holds 10 slides, 300/cs
ThermoSafe 369 Microscope Slide Mailer, holds 10 slides, 300/cs
0375010

$428.39CAD / Case of 300
+
ThermoSafe 365 Shipping Container/Carton for 03727-01
0372723

$294.31CAD / Case of 300
=
Total Price 722.70 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

More About this Item

Protect your fragile samples with this expanded polystyrene shipper. Light weight minimizes postage costs. Strong white paperboard shipping carton (sold separately) has sure-lock tabs that eliminate taping or tying. Choose from tube mailers, slide mailer, mini mailer, multipurpose mailer, or aluminum shipping can.

Specifications
  • Capacity10 slides
  • Outside Width (in)2 1/2
  • Outside Length (in)4
  • Outside Height (in)2
  • Outside Width (cm)6.4
  • Outside Length (cm)10.2
  • Outside Height (cm)5.1
  • Shipping Weight (oz)139
  • Shipping Weight (g)3946
  • DescriptionMicroscope Slide Mailer, 10 slides, 1" x 3"; 300/cs
Reviews
Q&A