ThermoSafe 361UPS Boston Round Bottle Shipper, Horizontal, 16 oz/473 mL; 64/Cs

Representative image only.

ThermoSafe 361UPS Boston Round Bottle Shipper, Horizontal, 16 oz/473 mL; 64/Cs

ThermoSafe | Mfr: 361UPS| Item: 0372935

Economical, expanded polystyrene shippers (EPS) for nonhazardous liquids

$283.26CAD / Case of 64

$283.26CAD / Case of 64
Product Description

More About this Item

A fast, cost-effective way to pack and transport bottles—just tape, label, and ship. Bottles are not included.

Specifications
  • Bottle Capacity (mL)473
  • Inside Height (in)7 1/2
  • Outside Width (in)4 1/2
  • Outside Length (in)8 9/16
  • Outside Height (in)4 1/2
  • Inside Diameter (ID) (in)3 1/4
  • Inside Height (cm)19.1
  • Outside Width (cm)11.4
  • Outside Length (cm)21.7
  • Outside Height (cm)11.4
  • Inside Diameter (ID) (cm)8.3
  • Shipping Weight (oz)192
  • Shipping Weight (g)5443
  • DescriptionBoston Round Bottle Shipper, Horizontal, 16 oz/473 mL; 64/Cs
Reviews
Q&A