ThermoSafe 344 Boston Round Bottle Shipper, Vertical, 8 oz/236 mL; 120/Cs

Representative image only.

ThermoSafe 344 Boston Round Bottle Shipper, Vertical, 8 oz/236 mL; 120/Cs

ThermoSafe | Mfr: 344| Item: 0372925

Economical, expanded polystyrene shippers (EPS) for nonhazardous liquids

$331.51CAD / Case of 120

$331.51CAD / Case of 120
Product Description

More About this Item

A fast, cost-effective way to pack and transport bottles—just tape, label, and ship. Bottles are not included.

Specifications
  • Bottle Capacity (mL)236
  • Inside Height (in)5 5/8
  • Outside Width (in)4
  • Outside Length (in)7
  • Outside Height (in)4
  • Inside Diameter (ID) (in)2 3/8
  • Inside Height (cm)14.3
  • Outside Width (cm)10.2
  • Outside Length (cm)17.8
  • Outside Height (cm)10.2
  • Inside Diameter (ID) (cm)6
  • Shipping Weight (oz)192
  • Shipping Weight (g)5443
  • DescriptionBoston Round Bottle Shipper, Vertical, 8 oz/236 mL; 120/Cs
Reviews
Q&A