ThermoSafe 341 Shipper, 8 Tube Mailer, 16 x 103mm tubes, 50/cs

Representative image only.

ThermoSafe 341 Shipper, 8 Tube Mailer, 16 x 103mm tubes, 50/cs

ThermoSafe | Mfr: 341| Item: 0372703

Ship non-hazardous specimens safely

$139.24CAD / Case of 50

$139.24CAD / Case of 50

Buy It Together

Current Item

ThermoSafe 341 Shipper, 8 Tube Mailer, 16 x 103mm tubes, 50/cs
ThermoSafe 341 Shipper, 8 Tube Mailer, 16 x 103mm tubes, 50/cs
0372703

$139.24CAD / Case of 50
+
ThermoSafe 365 Shipping Container/Carton for 03727-01
0372723

$294.31CAD / Case of 300
=
Total Price 433.55 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

More About this Item

Protect your fragile samples with this expanded polystyrene shipper. Light weight minimizes postage costs. Strong white paperboard shipping carton (sold separately) has sure-lock tabs that eliminate taping or tying. Choose from tube mailers, slide mailer, mini mailer, multipurpose mailer, or aluminum shipping can.

Specifications
  • CapacityEight 16 x 103 mm tubes
  • Inside Height (in)1/4
  • Inside Length (in)1 1/2
  • Inside Width (in)1
  • Outside Width (in)5 1/2
  • Outside Length (in)10
  • Outside Depth (in)1 3/4
  • Inside Height (cm)0.6
  • Inside Length (cm)3.8
  • Inside Width (cm)2.5
  • Outside Width (cm)14
  • Outside Length (cm)25.4
  • Outside Depth (cm)4.4
  • Shipping Weight (oz)89.6
  • Shipping Weight (g)2540
  • DescriptionShipper, 8 Tubes, 16 x 103mm tubes, 50/cs
Reviews
Q&A