ThermoSafe 338UPS Cider Jug Shipper, Vertical, 128 oz/3785 mL; 8/Cs

Representative image only.

ThermoSafe 338UPS Cider Jug Shipper, Vertical, 128 oz/3785 mL; 8/Cs

ThermoSafe | Mfr: 338UPS| Item: 0372942

Economical, expanded polystyrene shippers (EPS) for nonhazardous liquids

$134.67CAD / Case of 8

$134.67CAD / Case of 8
Product Description

More About this Item

A fast, cost-effective way to pack and transport bottles—just tape, label, and ship. Bottles are not included.

Specifications
  • Bottle Capacity (mL)3785
  • Inside Height (in)11 1/8
  • Outside Width (in)10
  • Outside Length (in)10
  • Outside Height (in)14 5/8
  • Inside Diameter (ID) (in)6 1/4
  • Inside Height (cm)25.3
  • Outside Width (cm)25.4
  • Outside Length (cm)25.4
  • Outside Height (cm)37.1
  • Inside Diameter (ID) (cm)15.9
  • Shipping Weight (oz)192
  • Shipping Weight (g)5443
  • DescriptionCider Jug Shipper, Vertical, 128 oz/3785 mL; 8/Cs
Reviews
Q&A