Thermo Scientific VSTAR52 Versa Star with pH/Conductivity Modules Kit

Representative image only.

Thermo Scientific VSTAR52 Versa Star with pH/Conductivity Modules Kit

Thermo Scientific | Mfr: VSTAR52| Item: 5882592

Modularity allows you to configure the meter how you need it--for now and for later

$6,014.97CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$6,014.97CAD / Each

Buy It Together

Current Item

Thermo Scientific VSTAR52 Versa Star with pH/Conductivity Modules Kit
Thermo Scientific VSTAR52 Versa Star with pH/Conductivity Modules Kit
5882592

$6,014.97CAD / Each
+ =
Total Price 6288.65 ADD 2 ITEMS TO CART
Product Description

Includes

VERSA STAR meters include: an electrode arm with redesigned holder, and a universal power adapter.

Key Features

  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Selectable cell constant for 2- or 4-cell conductivity probes
  • Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
  • Choice of practical salinity or natural sea water curves for salinity
  • 3 year warranty

More About this Item

The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.

This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.

Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.

Kit Adds one pH and one Conductivity measuring modules, ROSS pH Triode probe(05718-28), DuraProbe conductivity cell (19704-70), ROSS solution kit(475-mL of pH pH 4,7,10 buffers, storage solution and cleaning solution), Five x 60-mL bottles of 1413µS standard.

3 Year WarrantyIP54 Rated
Specifications
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Conductivity Accuracy±0.5% FS +1 LSD
  • Min TDS0 to 200 ppt
  • Min Salinity0 to 80 ppt
  • Salinity Accuracy±0.1 ppt
  • Resistivity Accuracy±0.5% ±1 digit
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)90 to 260
  • Power (Hz)50 or 60
  • Battery4 AA (not included)
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionVersa Star with pH/Conductivity Modules Kit
  • Warranty3 year
Reviews
Q&A