Thermo Scientific VSTAR40A2 Versa Star with pH/ISE Module Kit

Representative image only.

Thermo Scientific VSTAR40A2 Versa Star with pH/ISE Module Kit

Thermo Scientific | Mfr: VSTAR40A2| Item: 5882588

Modularity allows you to configure the meter how you need it--for now and for later

$6,664.73CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$6,664.73CAD / Each

Buy It Together

Current Item

Thermo Scientific VSTAR40A2 Versa Star with pH/ISE Module Kit
Thermo Scientific VSTAR40A2 Versa Star with pH/ISE Module Kit
5882588

$6,664.73CAD / Each
+ =
Total Price 6938.41 ADD 2 ITEMS TO CART
Product Description

Includes

VERSA STAR meters include: an electrode arm with redesigned holder, and a universal power adapter.

Key Features

  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included
  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Advanced ISE features such as auto-blank, low-concentration stability range, and incremental techniques
  • 3 year warranty

More About this Item

The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.

This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.

Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.

Kit adds one pH/ISE/Temperature measuring module, ROSS Ultra pH electrode (05718-28), ATC probe (58824-45), ROSS Solution kit(475-mL bottle of pH 4, 7, 10 buffers; storage solution and cleaning solution).

3 Year WarrantyIP54 Rated
Specifications
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.05% of reading
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)90 to 260
  • Power (Hz)50 or 60
  • Battery4 AA (not included)
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionVersa Star with pH/ISE Module Kit
  • Warranty3 year
Reviews
Q&A