Thermo Scientific STAR A329 Star A329pH/ISE/Cond/DO Portable Meter Kit

Representative image only.

Thermo Scientific STAR A329 Star A329pH/ISE/Cond/DO Portable Meter Kit

Thermo Scientific | Mfr: STARA3295| Item: 5882578

Advanced features and dependability makes it the top choice for measuring samples

$5,615.83CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$5,615.83CAD / Each

Buy It Together

Current Item

Thermo Scientific STAR A329 Star A329pH/ISE/Cond/DO Portable Meter Kit
Thermo Scientific STAR A329 Star A329pH/ISE/Cond/DO Portable Meter Kit
5882578

$5,615.83CAD / Each
+ =
Total Price 5889.51 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A 300 meters features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

GLP-related features include 5000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Available as meter only and as complete meter kit configurations

Kit includes ROSS pH/ATC electrode, Conductivity probe, RDO optical probe, 10 pouches each of 4, 7, 10 buffer, 1413µS standard, and rinse solution, protective armor with probe holders, hard carrying case, and four AA batteries.

3 Year WarrantyIP 67
Specifications
  • Meter TypePortable
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.2 or 0.05% of reading
  • Min Conductivity0.001 μS to 3000 mS
  • Conductivity Accuracy±0.5% FS + 1 LSD
  • Min TDS0 to 200 ppt
  • Min Salinity0 to 80 ppt
  • Salinity Accuracy±0.1 ppt
  • Resistivity Resolution2 Ω
  • Resistivity Accuracy±0.5% ±1 digit
  • Min Dissolved Oxygen0 to 90 ppm; 0 to 600 %
  • Dissolved Oxygen Accuracy±0.2 ppm
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)10.795
  • Height (cm)23.1775
  • Width (in)4 1/4
  • Length (in)2 3/8
  • Height (in)9 1/8
  • Probe IncludedYes
  • DescriptionStar A329pH/ISE/Cond/DO Portable Meter Kit
  • Warranty3 year
Reviews
Q&A